Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ DTT (Dithiothreitol), No-Weigh™ Format

Catalog No. pia39255
Click to view available options
Quantity:
48 x 7.7 mg

Thermo Scientific™ Pierce™ DTT (Dithiothreitol) No-Weigh™ Format is designed for convenient preparation of stocks that will be added to protein samples for effective reduction of disulfide bonds.

Thermo Scientific™ Pierce DTT (Dithiothreitol) is offered as 7.7 mg x 48-tube microplates for convenient preparation of 100 μL of 0.5 M stocks for addition to protein samples for effective reduction of disulfide bonds.

Thermo Scientific Pierce DTT (Dithiothreitol), No-Weigh format, is offered as 7.7 mg x 48-tube microplates for convenient preparation of 100 μL of 0.5 M single-use stocks for addition to protein samples for effective reduction of disulfide bonds.

DTT is a popular reducing agent for many protein research applications, including preparing proteins for reducing polyacrylamide electrophoresis (SDS-PAGE) and reducing disulfide bonds for labeling sulfhydryl groups in conjugation reactions. At low concentrations, DTT is used to stabilize proteins and enzymes with free sulfhydryl groups to keep monothiols in a reduced state. DTT reduction is also used to prepare proteins for mass spectrometry.

Properties of DTT:

Synonyms—1,4-dithiothreitol; DTT; Cleland’s Reagent; 1,4-dimercapto-2,3-butanediol
Molecular weight (MW)—154.24
Molecular formula—C4H10O2S2
CAS number—3483-12-3

DTT is not stable in solution and should be prepared fresh for each use. The No-Weigh format consists of a 48-microtube tray containing dry, stable, single-use aliquots of the DTT chemical to eliminate the need to weigh small amounts of reagent and prevent waste of unused DTT. Just puncture the seal of one microtube, add 100 μL of water or buffer, and mix to dissolve. In just a few seconds you will have a solution of 500 mM DTT, ready for a variety of laboratory methods. Dilute as necessary or add the appropriate volume of the 500 mM stock solution to achieve the amount of DTT reducing agent required for your specific application.

Features of the No-Weigh format:

Saves time—just add 100 μL water or buffer to make 500 mM DTT solution in seconds
Eliminates waste—makes 100 μL at a time, just the right amount for most applications
Always fresh—ensures full reducing strength for every use; no need to store stock solutions

With the convenient No-Weigh format, a 500 mM DTT solution can be made in less than 30 seconds. The unique packaging ensures that the DTT reducing agent is at full strength and able to protect proteins from oxidative damage or reduce any disulfides prior to electrophoresis.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 48 x 7.7 mg
Content And Storage Upon receipt, store product at 4°C.

Product Line Pierce™

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.