Learn More
Thermo Scientific™ Pierce™ EasySep Human CD8+ T Cell Enrichment Kit
Catalog No. ENCS005
Change view
Click to view available options
:
Human CD8+ T cell
Catalog No.
ENCS005 Human CD8+ T cell
1 options
Catalog No. ENCS005
Supplier: Thermo Scientific™ CS005
Enriches CD8+T cells from fresh or previously frozen peripheral blood mononuclear cells by depletion of non-CD8+T cells
  • The kit labels 1 x 109 non-CD8+T cells for magnetic separation
  • Uses a cocktail of purified monoclonal antibodies bound in bispecific Tetrameric Antibody Complexes (TAC) directed against cell surface antigens on human blood cells (CD4, CD14, CD16, CD19, CD56, TCR gamma/delta, glycophorin A) and dextran
  • Purity: 83 - 91%
  • Recovery: 46 - 60%