Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ Endotoxin-Free Water PROMO
Click to view available options
:
Thermo Scientific™ Pierce™ Endotoxin-Free Water
Description
Pierce Endotoxin-Free Water is free of endotoxins and enzymes, including proteases. Using Endotoxin-Free Water helps eliminate the risk of harmful activity produced by bacteria that can interfere with cell health and function. It is suitable for usage in molecular biology or general biochemistry applications and contains <0.005 EU/mL endotoxin.
Specifications
Specifications
Endotoxin | <0.005 EU/mL |
Packaging | Bottle |
Sterility | Sterile |
Quantity | 4 x 50 mL |
Purification Method | Sterile filtered with 0.5 μm filter |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction