Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ F(ab')2 Micro Preparation Kit PROMO

Catalog No. PI44688PM
Change view
Click to view available options
:
Thermo Scientific™ Pierce™ F(ab')2 Micro Preparation Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PI44688PM Thermo Scientific™ Pierce™ F(ab')2 Micro Preparation Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI44688PM Supplier Thermo Scientific™ Supplier No. 44688

Selectively digest whole human and other IgG antibodies with immobilized pepsin protease to make F(ab')2 fragments that retain antigen binding activity.

These kits use pepsin, a nonspecific endopeptidase, to enzymatically digest the Fc portion of whole IgG to yield the fragment known as F(ab')2. This fragment is composed of a pair of Fab' units connected by two disulfide bonds. Because the pepsin protease is supplied in immobilized form as beaded agarose resin, the digestion reaction is easily stopped by removing the resin from the IgG solution; the result is digest products that are enzyme-free. An optimized procedure, high-performance desalting and Protein A spin columns make these kits easy to use.

Highlights:

  • Enzyme-free digestion products – immobilized pepsin (beaded agarose resin) provides for control of the digestion reaction and complete removal of resulting antibody fragments from the protease

  • Suitable for human and other species of IgG – the kit procedure is optimized for human, rabbit and mouse IgG, but pepsin-based digestion is effective for other species and subclasses of IgG (Note: for best results with mouse IgG1, use Part No. 44980)

  • High capacity – kit provides for digestion and purification from as much as 4mg of IgG at a time. Use the standard size kit for fragmenting 0.25 to 4mg IgG, and the micro kit for 25 to 250μg IgG

  • Complete and easy to use – kits include all reagents, spin columns and accessories needed to prepare and purify antibody fragments

Includes:

Pepsin agarose resin, digestion buffer, Protein A columns with binding and elution buffers, spin desalting columns and accessories

Specifications

Product Line Pierce
Quantity 10 Antibody Samples, each containing 25 to 250 μg IgG
Content And Storage Sufficient For: 10 antibody samples, each containing 25 to 250 μg IgG, Pierce Pepsin Agarose, 0.5 mL, F(ab')2 Digestion Buffer, 55 mL, NAb Protein A Plus Spin Columns, 0.2 mL, 2 Columns, PBS Packs (each makes 500 mL), 2 packs, IgG Elution Buffer, 50 mL, Zeba Spin Desalting Columns, 7K MWCO, 0.5 mL, 10 Columns, Spin Columns, 0.8 mL, 10 Columns, Microcentrifuge Tubes, 2 mL, 30 Tubes, Store at 4°C
Target Gene ERK5/BMK1 (pan)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.