Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Firefly Luciferase Flash Assay Kit

Catalog No. p-4532171
Click to view available options
Quantity:
100 Reaction Kit
1000 Reaction Kit

Assay regulatory element transcriptional activity in whole cell lysates with this highly sensitive flash-based firefly luciferase kit.

The Thermo Scientific™ Pierce Firefly Luciferase Flash Assay Kit provides reagents to measure Red firefly luciferase activity in whole cell producing a very strong flash signal which decays rapidly.

Thermo Scientific Pierce Firefly Luciferase Flash Assay Kit contains reagents to measure the activity of firefly luciferase in mammalian whole cell lysates. Containing a convenient lysis buffer, assay buffer and firefly substrate, this kit provides a complete reagent system for the intracellular detection of promoter or pathway activity. Light output resulting from the luciferase reaction is captured using a luminometer and can be correlated with the amount of firefly luciferase protein produced and used to determine the activity of the promoter driving firefly expression. These reagents are optimized to work with Thermo Scientific Firefly Luc Plasmids; however, the Flash Assay Kit may be used to detect the activity of other firefly or ATP-dependent luciferases utilizing D-luciferin as a substrate.

Highlights:

  • Highly sensitive detection of firefly luciferase activity
  • Use with Thermo Scientific Pierce Firefly Luciferase Signal Enhancer for best results (maximum signal)
  • Working solution stable for up to 2 months at -20°C
  • Assay reagents compatible with other firefly luciferases
  • Amenable to automation
  • Contains a universal cell lysis buffer and optimized flash assay reagent
  • Safe non-radioactive assay

Includes:

Cell lysis buffer, reaction buffer and substrate

Requires:

Firefly luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers; For best results (maximum signal), use with Thermo Scientific Pierce Firefly Luciferase Signal Enhancer

Recommended for:

Promoter studies for analyzing cis- regulatory elements and trans-acting factors; Drug screening; siRNA and miRNA screening; Multiplexed assays to study off-target effects; Secretory pathway / protein localization reporter assays; Signal transduction pathway analysis; RNA splicing studies

TRUSTED_SUSTAINABILITY

Specifications

Type Assay Kit
Target Luciferase, Firefly Luciferase
Quantity 100 Reaction Kit
Content And Storage • Firefly Flash Assay Buffer, 5 mL, store at 4°C
• D-Luciferin, Lyophilized, 3 mg, store at 4°C
• 2X Cell Lysis Buffer, 6 mL, store at room temperature

Upon receipt store kit at 4°C or store individual components as indicated above.
Assay Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells Mammalian Cells
Detection Method Bioluminescence
For Use With (Equipment) Luminometer (Microplate)
Format 384-well plate, 96-well plate
Label Type Enzyme Labeled
Substrate Properties Chemical Substrate
Technique Enhanced Chemiluminescence
Substrate Type Luciferase Substrate
Product Line Pierce
Substrate D-Luciferin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.