Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ FlexMix™ Calibration Solution PROMO
Click to view available options
:
Thermo Scientific™ Pierce™ FlexMix™ Calibration Solution
Description
- Improved mass accuracy - extended mass range improves mass accuracy
- Positive and negative mode calibration - single formulation with extended mass ranges for both ionization calibration modes
- Highly validated - high-quality, rigorously-tested formulation with lot-specific certificates of analysis
- Ready-to-use - reference standard may be directly loaded into a syringe and injected into the instrument
- Store at room temperature for up to 18 months
Specifications
Specifications
For Use With (Equipment) | Mass Spectrometer |
Product Type | Calibration Solution |
Chemical Reactivity | Amine |
Product Line | Pierce™ |
Shipping Condition | Room Temperature |
Workflow Step | Calibration |
Quantity | 10 mL |
Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction