Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Gaussia-Firefly Luciferase Dual Assay Kit

Catalog No. p-4532163
Click to view available options
Quantity:
100 Reaction Kit
1000 Reaction Kit

Simultaneously detect intracellular Gaussia and Red Firefly luciferase activity in mammalian whole cell lysates.

The Thermo Scientific™ Pierce Gaussia-Firefly Luciferase Dual Assay Kit provides the necessary reagents to simultaneously detect intracellular Gaussia and Red Firefly luciferase activity in mammalian whole cell lysates.

The Pierce Gaussia-Firefly Luciferase Dual Assay Kit is a highly sensitive assay that allows for the simultaneous detection of Gaussia and Red Firefly luciferase activity. Gaussia luciferase acts as an experimental reporter with constitutively active red firefly as a normalization control. This reporter-and-control combination enables simultaneous monitoring of experimental reporter and control luciferase activities in a single-read assay without the need for two-step addition of substrate reagents or quenching. The assay working solution contains substrates for both luciferases, and the reactions occur simultaneously with flash-type kinetics. The resulting luminescent signals are spectrally resolvable using filters. In a single sample, researchers can assay transcriptional activity of regulatory elements, signal transduction pathways, and effects of activators or inhibitors.

Highlights:

  • Simultaneous – concurrent, filter-based, wavelength-separated detection of two luciferase activities

  • Sensitive – measure bright blue Gaussia and red firefly luciferase activities in the same sample

  • Fast – no quenching step required, unlike in traditional sequential dual assays

  • Multiplex – capable of quantitating two cellular activities in the same sample(s)

Includes:

Cell lysis buffer, reaction buffer, and substrates coelenterazine and D-luciferin

Requires:

Gaussia and Red Firefly luciferase reporters; Filter sets in the ranges of 425 to 525nm for Gaussia Luc and 615 to 675nm for Red Firefly Luc. Luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers; Injector required for assessing more than 24 wells at a time.

Recommended for:

Study two regulatory elements at the same time; Monitor two signaling pathways simultaneously; Enable studying more than one target per screen (e.g. off-target effects)

TRUSTED_SUSTAINABILITY

Specifications

Type Assay Kit
Target Luciferase, Firefly Luciferase, Gaussia Luciferase
Quantity 100 Reaction Kit
Content And Storage • Gaussia-Firefly Dual Assay Buffer, 5 mL, store at 4°C
• Coelenterazine (100X), 50 μL, store at -80°C
• D-Luciferin, Lyophilized, 3 mg, store at 4°C
• 2X Cell Lysis Buffer, 6 mL, store at room temperature

Upon receipt store kit at -80°C or store individual components as indicated above.
Assay Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells Mammalian Cells
Detection Method Bioluminescence
For Use With (Equipment) Luminometer (Microplate)
Format 384-well plate, 96-well plate
Label Type Enzyme Labeled
Substrate Properties Chemical Substrate
Technique Enhanced Chemiluminescence
Substrate Type Luciferase Substrate
Product Line Pierce™
Substrate Coelenterazine, D-Luciferin
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.