Learn More
Thermo Scientific™ Pierce™ β-Glucan Blocker PROMO
Description
Pierce β-Glucan-Blocker is added to samples suspected of β-1,3-glucans contamination, preventing the occurrence of false positive results in endotoxin detection (LAL) assays. β-1,3-glucans contamination may arise from the presence of yeast and cellulosic materials. β-1,3-glucans, in sufficient amounts, can trigger a LAL response not emanating from an endotoxin-mediated response. This blocker contains <0.005 EU/mL endotoxin.
Specifications
Specifications
| Content And Storage | Store at 2 to 30°C. |
| Product Type | Beta-Glucans Blocker |
| Sterility | Sterile |
| Endotoxin Level | <0.005 EU/mL |
| Format | Vial(s) |
| For Use With (Application) | LAL Endotoxin Assay |
| Quantity | 5 x 10 mL |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.