Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ GPCR Extraction and Stabilization Reagent PROMO
Click to view available options
:
Thermo Scientific™ Pierce™ GPCR Extraction and Stabilization Reagent
Description
- Thermo Scientific GPCR Extraction and Stabilization Reagent enables efficient extraction and stabilization of G-coupled protein receptors (GPCRs) and other membrane-associated proteins from cultured mammalian cells or tissue while preserving longer-term receptor integrity.
- Ease-of-use—all-in-one formulation to extract and stabilize GPCRs in one hour
- GPCR preservation—stabilizes the receptor in a detergent micelle to ensure receptor function
- Optimized—formulation and procedure provide gentle but efficient extraction of GPCRs from the plasma membrane
- Compatible—lysate is directly compatible with downstream applications including western blotting, immunoprecipitation, and receptor ligand binding assays
- GPCR Extraction and Stabilization Reagent is simple to use. The sample is first harvested, washed, and pelleted. Receptor is then extracted using the reagent, followed by centrifugation. The resulting whole cell lysate contains solubilized and stabilized receptors. The reagent has been tested for use with endogenous and overexpressed GPCRs, and receptor function has been verified by receptor ligand binding assays.
- GPCRs are a large family of receptors with seven transmembrane domains that play a key role in transmitting signals across the cell membrane. They are effective drug targets for genetic and immune system disorders and serve as potential biomarkers and therapeutic targets for cancer treatment. One of the major challenges in GPCR research is the ability to efficiently extract the receptor from the membrane without compromising the structural and functional integrity of the protein.
- GPCR Extraction and Stabilization Reagent encapsulates the receptor in a detergent micelle optimized to ensure receptor function for up to one week when stored at 4°C and one month when stored at -20°C post extraction, providing more flexibility for receptor analysis.
Specifications
Specifications
Quantity | 100 mL |
Format | Liquid |
Product Type | Reagent |
Volume (Metric) | 100 mL |
Content And Storage | Store in refrigerator (2°C to 8°C). |
Shipping Condition | Room Temperature |
Product Line | Pierce™ |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction