Learn More
Thermo Scientific™ Pierce™ GST Agarose PROMO
Description
This GST resin consists of purified glutathione S-transferase protein from Schistosoma japonicum that has been immobilized on crosslinked 6% beaded agarose. Antibody molecules in that cross-react with the GST portion of a recombinant fusion protein can be removed from the desired protein-specific antibody by passing antiserum over a GST agarose column. GST antibodies will bind to the immobilized GST, whereas antibodies against other fusion proteins will pass through the column unrestrained.
Highlights:
Specific – provides purification of target antibodies from those directed against the GST tag
Reusable – bound antibodies can be eluted with 6M guanidinium chloride allowing for 10 purifications with each column or aliquot of resin
Two formats – choose either pre-packed columns or resin slurry for use at a variety of scales
Specifications
Specifications
Product Type | Agarose |
Product Line | Pierce™ |
Column Type | Agarose Resin, Affinity |
Format | Bottle |
Stationary Phase | GST Protein |
Purification Target | Anti-GST Antibody |
Content And Storage | 4°C |
Quantity | 5 mL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.