Learn More
Thermo Scientific™ Pierce™ HA-Tag IP/Co-IP Kit

Description
Thermo Scientific™ Pierce HA Tag IP and Co-IP Kit provides the affinity resin and other reagents necessary to easily perform immunoprecipitation (IP) or co-immunoprecipitation (co-IP) reactions using an HA-tagged protein as the bait.
With sufficient materials to perform 25 IP or Co-IP assays, the complete HA Tag IP/Co-IP Kit includes the immobilized anti-HA antibody on agarose resin, binding, wash and elution buffers, microcentrifuge spin columns, HA-tag positive control lysate, sample loading buffer and easy-to-follow instructions.
Highlights:
- Improved updated kit includes more immobilized antibody resin and improved protocol along with GST-PI3K-SH2-HA fusion protein as a positive control
- Specific highly specific anti-HA monoclonal antibody provides high-yield immunoprecipitation products and clean Western blot detection
- Compatible includes protocols and reagents for multiple elution conditions to accommodate different protein sensitivities and downstream applications
- Robust the kit and the anti-HA agarose are compatible with various cell lysates and physiologic (non-denaturing) buffer systems
- Convenient and Easy complete kit includes all necessary reagents, convenient spin columns and easy-to-follow instructions
Includes:
Anti-HA agarose resin, TBS pack, elution buffer, lane marker nonreducing sample buffer, positive control lysate, microcentrifuge spin columns and collection tubes
Recommended for:
Purification and detection of HA-tagged fusion-protein for downstream assays; Enrichment and screening of HA-tagged fusion-protein mutants; Co-immunoprecipitation and screening of interacting proteins
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Content And Storage | Sufficient For: 25 IP/Co-IP reactions, each using 10 μL of resin • Anti-HA Agarose, 0.25 mL • HA-tagged Positive Control, 0.5 mL • BupH Tris Buffered Saline Pack (500 mL), 1 pack • Lane Marker Non-Reducing Sample Buffer (5X), 5 mL • Pierce Spin Columns Accessory Pack, 27 columns • Collection Tubes and Caps Accessory Pack, 100 tubes Store HA-tagged positive control at -80°C (may be stored at -20°C for up to 6 months). Store remaining kit components at 4°C. |
Assay | Affinity Purification |
For Use With (Equipment) | Microcentrifuge |
Product Line | Pierce™ |
Quantity | 25 reactions kit |
For Use With (Application) | Pull-Down (Tag-Based), Immunoprecipitation (IP) |
Target | Target-Specific |
Type | Tag IP/Co-IP Kit |
Sufficient For | 25 Reactions |
Format | Kit |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.