Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ HA-Tag Magnetic IP/Co-IP Kit
GSA_VA

Catalog No. PI88838
Click to view available options
Quantity:
40 reactions

Perform immunoprecipitation (IP) assays of HA-tagged proteins or co-IP reactions with HA-tagged bait proteins with these magnetic beads, positive control and reagents.

Perform immunoprecipitation (IP) assays of HA-tagged proteins or co-IP reactions with HA-tagged bait proteins with the magnetic beads, positive control and reagents provided in the Thermo Scientific™ Pierce Magnetic IP/Co-IP Kit.

Additional Information and Online Ordering

The complete Magnetic IP/Co-IP Kit can be used to perform 40 IP or Co-IP assays. It includes magnetic beads coated with immobilized anti-HA antibody, lysis/wash buffer, low pH elution buffer, neutralization buffer, HA-tag positive control lysate, and non-reducing sample buffer. Protocols are provided for both manual and automated workflows.

Highlights:

  • Specific – highly specific anti-HA monoclonal antibody enables high yields of immunoprecipitation products
  • Fast – Optimized protocol is completed in approximately 1 hour
  • Low non-specific binding – stable, pre-blocked beads provide highly purified HA-tagged proteins
  • Versatile – beads are compatible with manual and automated workflows (e.g., Thermo Scientific™ KingFisher Instruments)
  • Convenient and easy – complete kit includes all necessary reagents and easy-to-follow instructions

Includes:

The kit contains magnetic beads immobilized with anti-HA antibody, lysis/wash buffer, low pH elution buffer, neutralization buffer, HA-tag positive control lysate, and non-reducing sample buffer.

Recommended for:

High-throughput immunoprecipitation of recombinant HA-tagged proteins and co-IP of interacting proteins

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1 mL
Formulation Slurry: 10 mg/mL, 1% solids
Content And Storage Sufficient For: 40 IP reactions using 25 μL of anti-HA magnetic beads
• HA-tagged Positive Control (Part No. 26180X), 500 μL
• Pierce Anti-HA Magnetic Beads, 1 mL
• Pierce IP Lysis/Wash Buffer, 2 x 50 mL
• Non-Reducing Sample Buffer (5X), 5 mL
• Elution Buffer (pH 2.0), 5 mL
• Neutralization Buffer (pH 8.5), 1 mL

HA-tagged Positive Control ships separately—store at -70°C. Store remaining components at 4°C.
Assay Affinity Purification
For Use With (Equipment) KingFisher™ Flex, Magnetic Stand
Product Line Pierce™
Quantity 40 reactions
For Use With (Application) Pull-Down (Tag-Based), Immunoprecipitation (IP)
Target Target-Specific
Type Tag IP/Co-IP Kit
Sufficient For 40 Reactions
Format Kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.