Learn More
Thermo Scientific™ Pierce™ HeLa Digest/PRTC Standard, 10 μg PROMO
Description
The Thermo Scientific Pierce HeLa Digest/PRTC Standard is a validated complex mammalian protein digest containing additional PRTC heavy isotope-labeled peptides designed for LC/MS performance testing.
Features of HeLa Digest/PRTC Standard include:
- Convenient—pre-mixed formulation of HeLa digest and PRTC standard in a ready-to-use format
- Informative—enables robust performance assessment of LC/MS instruments
- Verified—high-quality, consistent mixture documented via lot-specific certificates of analysis
- Stable—provided in a lyophilized format
The Pierce HeLa Digest/PRTC Standard is a lyophilized mixture of 10 μg of HeLa protein digest with 5 pmol PRTC peptides to assess chromatography MS instrument performance. The protein digest is derived from a well-established adenocarcinoma reference HeLa S3 cell line that expresses over 15,000 proteins with relevant post-translational modifications. This complex digest is an ideal standard for complex proteome mass spectrometry applications.
The included Pierce Peptide Retention Time Calibration Mixture contains 15 synthetic heavy isotope-labeled peptides mixed at an equimolar ratio that elute across the chromatographic gradient. The observed retention times and hydrophobicity factors (HFs) for these calibrant peptides are fit to a linear equation to determine the slope of the retention time/HF relationship. This equation and the HF of an uncharacterized peptide are then used to predict retention time. In combination, the mixture is a powerful tool to optimize and assess LC parameters, identify the total peptide elution window, and optimize and monitor MS instrumentation.
This standard is specifically formulated for LC/MS experiments and does not contain salts or detergents. Using the standard routinely before analysis of complex samples makes it possible to monitor and normalize LC/MS performance between samples and over time.
Applications:
- Qualitative LC assessment
- QC assay tool for assessment of LC/MS system performance
- LC/MS standardization
- LC/MS method development and optimization
- Normalize results for variation in retention times and peak intensities between runs
Specifications
Specifications
| Chemical Name or Material | HeLa Digest/PRTC Standard |
| Packaging | Glass Vial |
| Physical Form | Lyophilized |
| Synonym | HeLa/PRTC |
| Quantity | 10 μg |
| Grade | MS |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.