Learn More
Thermo Scientific™ Pierce™ High Capacity Endotoxin Removal Resin, 100 mL PROMO
Description
The Pierce High Capacity Endotoxin Removal Resin combines porous cellulose beads and an FDA-approved food preservative, poly(epsilon-lysine), as an affinity ligand to selectively bind endotoxins. Endotoxin levels in protein samples are reduced by greater than 99% in as little as 1 hour using our spin cup format, and protein recovery is ≥85%. The modified polylysine affinity ligand eliminates the toxicity concerns associated with alternative technologies using polymixin B ligands and sodium deoxycholate buffers. Pierce High capacity Endotoxin Removal Resin is available as a slurry to pack a custom column that can be re-used at least 5 times with no loss in endotoxin removal efficiency. This resin is also available in a new single-use spin column format optimized for different sample volumes.
Highlights:
High capacity bind up to 2,000,000 EU/mL to eliminate at least 99% of endotoxins
Durable re-use resin up to 10 times with no loss in performance
Selective recover at least 85% of your protein sample
High performance compatible with FDA guidelines by reducing final EU concentration to less than 5 EU/mL
Fast our new spin cup format enables endotoxin depletion within 1 hour
Cleansingle-use spin columns avoid cross contamination of samples
Optimized spin cups are optimized for different sample volumes
Economical large-volume discounts available
Requires:
Endotoxin-free water
Recommended for:
Eliminate endotoxins from protein or antibody samples prior to cell transfection or animal injection
Specifications
Specifications
Capacity (Metric) | Up to 2,000,000 EU/mL |
Formulation | Proprietary ligand on beaded cellulose slurried in 20% ethanol |
Content And Storage | Upon receipt store at 4°C. |
Form | Liquid |
Product Line | Pierce™ |
Quantity | 100 mL |
Target | Endotoxin Removal |
Type | Resin |
Stationary Phase | Proprietary Ligand |
Format | Bottle |
Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.