Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ High Capacity Protein A MagBeads, alkali stable, 25 mL PROMO
Click to view available options
:
Thermo Scientific™ Pierce™ High Capacity Protein A MagBeads
Description
- High-performance magnetic beads for purifying antibodies from cell culture supernatant, ascites, serum, and crude extracts
- The ligand is produced in yeast and is immobilized on magnetic agarose with an average particle size of ∼30 mm
- Engineered to withstand standard CIP procedures with 0.1M NaOH, enabling efficient cleaning of the beads for reuse
- Beads can be regenerated at least 10 times without sacrificing binding capacity
- Suitable for purifying mAbs and a subpopulation of scFvs and Fab fragments containing a VH3 domain
- Compatible with manual and automated workflows (e.g., KingFisher instruments)
- High-affinity Protein A is securely immobilized to magnetic agarose to prevent leaching of ligand, target antibody, or base support
- High binding capacity of;≥ 40 mg HIgG/mL settled beads
- Animal component-free raw materials
Specifications
Specifications
| Binding Capacity | ≥40 mg HIgG/mL settled beads |
| Content And Storage | Provided in the form of a 25% slurry. Upon receipt Store at 2°C to 8°C |
| Particle Size | ∼ 30 μm |
| Quantity | 25 mL |
| For Use With (Application) | Antibody Purification |
| Ligand Type | Alkaline Stable Protein A |
| Format | Bottle |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction