Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ High pH Reversed-Phase Peptide Fractionation Kit

Catalog No. pi84868
Click to view available options
Quantity:
12 Reactions

Increase protein identification from LC/MS analysis through orthogonal peptide fractionation of complex peptide samples using Thermo Scientific™ Pierce™ High pH Reversed-Phase Peptide Fractionation Kit.

In order to enable deep proteome sequencing, it is often necessary to reduce the sample complexity by fractionation in an orthogonal dimension prior to LC/MS analysis. The Pierce High pH Reversed-Phase Peptide Fractionation Kit utilizes high pH reversed-phase chromatography to separate peptides by hydrophobicity, and provides excellent orthogonality to low pH reversed-phase LC-MS gradients. The kit has been designed to improve protein identification through the use of a proprietary reversed-phase resin in an easy-to-use spin column format with a high-pH fractionation protocol.

Thermo Scientific Pierce High pH Reversed-Phase Peptide Fractionation Kit increases protein identification from LC/MS analysis through orthogonal peptide fractionation of complex peptide samples.

  • Easy-to-use - resin provided in single-use spin column format
  • Improved protein identifications - protein identifications increased by ≥50% when compared to unfractionated samples
  • Reproducible - elution profiles and fractional resolution vary by less than 20%
  • Optimized - robust procedure for maximal protein identification and peptide recovery while minimizing fractional overlap
  • Compatible - reagents validated with a variety of complex samples, including TMT™-labeled peptides
In order to enable deep proteome sequencing, it is often necessary to reduce the sample complexity by fractionation in an orthogonal dimension prior to LC/MS analysis. The Pierce High pH Reversed-Phase Peptide Fractionation Kit utilizes high pH reversed-phase chromatography to separate peptides by hydrophobicity and provides excellent orthogonality to low pH reversed-phase LC-MS gradients.
The kit has been designed to improve protein identification through the use of a proprietary reversed-phase resin in an easy-to-use spin column format with a high-pH fractionation protocol. In contrast to strong cation exchange (SCX) fractionation, the high-pH reversed-phase fractions do not require an additional desalting step before LC/MS analysis.
The High pH Reversed-phase Peptide Fractionation Kit includes a high pH buffer (0.1% triethylamine) and twelve spin columns containing pH-resistant reversed-phase resin. Each reversed-phase fractionation spin column enables fractionation of 10 to 100μg of peptide sample using a microcentrifuge.
Native, phosphorylated, Tandem Mass Tag™ (TMT)-labeled, and other complex peptide mixture samples can be fractionated using this kit. Combining the search results generated by the individual fractions helps improve protein sequence coverage and increases the number of identified proteins relative to unfractionated samples.
Applications:
  • Reducing sample complexity to identify targets of interest
  • Conducting systems biology studies
  • Reducing sample complexity to improve quantitation studies

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Microcentrifuge
Product Type Peptide Fractionation Kit
Product Line Pierce™
Content And Storage Reversed-phase fractionation spin columns, 12 columns
Triethylamine (0.1%), 100 mL

Store in refrigerator (2–8°C).
Starting Material Peptides, Protease-digested Protein
Final Product Type Peptides
Detection Method Mass Spectrometry
Format Kit
Workflow Step Fractionation
Quantity 12 Reactions
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.