Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Horseradish Peroxidase

Catalog No. p-4532211
Click to view available options
Quantity:
10 mg
100 mg

Purified horseradish peroxidase enzyme for use in activity assays and conjugation to antibodies for ELISA, Western blot and immunohistochemistry applications.

Thermo Scientific™ Pierce Horseradish Peroxidase (HRP) is purified horseradish peroxidase enzyme for use in activity assays and conjugation to antibodies for ELISA, Western blotting and immunohistochemistry applications.

Highlights:

  • Superior to alkaline phosphatase and β-galactosidase conjugates due to the higher specific enzyme activity
  • Small size (40kDa) allows excellent cellular penetration
  • Variety of substrates available
  • Ideal in blotting and cytochemistry applications
  • Used as the reporter enzyme for Thermo Scientific SuperSignal Chemiluminescent Western Blotting and ELISA Substrates

Note:

Porstmann, B., Porstmann, T., Nugel, E. and Evers, U. (1985). Which of the commonly used marker enzymes gives the best results in colorimetric and fluorimetric enzyme immunoassays: horseradish peroxidase, alkaline phosphatase, β-galactosidase, J. Immunol. Meth. 79, 27-37.

Wordinger, R.J., Miller, G.W. and Nicodemus, D.S. (1987). Manual of Immunoperoxidase Techniques, 2nd Edition. Chicago: American Society of Clinical Pathologists Press, pp. 23-24.

Yolken, R.H. (1982). Enzyme immunoassays for the detection of infectious antigens in body fluids: current limitations and future prospects. Rev. Infect. Dis. 4(1), 35-68.

Cordell, J.L., et al. (1984). Immunoenzymatic labeling of monoclonal antibodies using immune complexes of alkaline phosphatase and monoclonal anti-alkaline phosphatase (APAAP complexes). J. Histochem. Cytochem. 32, 219-229.

Passey, R.B., et al. (1977). Evaluation and comparison of 10 glucose methods and the reference method recommended in the proposed product class standard. Clin. Chem. 23(1), 131.

Hosoda, H., Takasaki, W., Tsukamoto, R. and Nambara, T. (1987). Sensitivity of steroid immunoassays. Comparison of alkaline phosphatase, β-galactosidase and horseradish peroxidase as labels in a colorimetric assay system. Chem. Pharm. Bull. 35, 3336-3342.

Samoszuk, M.K., et al. (1989). Antibody, Immunoconjugates and Radiopharmaceuticals 2, 37-46.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.
Form Powder
For Use With (Application) Western Blotting, ELISA, Immunohistochemistry
Product Line Pierce™
Label or Dye HRP (Horseradish Peroxidase)

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.