Learn More
Thermo Scientific™ Pierce™ HRV 3C Protease Solution Kit (2 units/μL), 10,000 U PROMO
Description
Pierce HRV 3C Protease is highly specific for the recognition sequence Leu-Glu-Val-Leu-Phe-Gln▾Gly-Pro and cleaves after the glutamine residue. The recombinant enzyme is engineered as a fusion protein with glutathione-S-transferase (GST) and polyhistidine (6xHis) tags, so that the protease can be easily removed after cleavage using a variety of solid supports with immobilized glutathione (GSH) or metal chelates, respectively. HRV3C is also active in a variety of commonly used buffers, providing further flexibility in experimental design.
Highlights:
Effective – HRV 3C is designed to remove purification tags from your protein of interest
High activity at 4°
C
– activity is ≥2000U/mg control protein; one unit will cleave 0.1 mg control protein at 4°C after 16h
Flexible – dual-tagged enzyme is compatible with various columns and buffers
Includes:
Kit contains protease, cleavage buffer and control protein.
Recommended for:
Removal of purification tags from fusion proteins
Specifications
Specifications
Content And Storage | Store at -20°C. |
Product Line | Pierce™ |
Form | Kit |
Quantity | 10,000 U |
Product Type | Protease |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.