Learn More
Thermo Scientific™ Pierce™ HRV 3C Protease Solution Kit (2 units/μL), 1000 U PROMO
Description
Pierce HRV 3C Protease is highly specific for the recognition sequence Leu-Glu-Val-Leu-Phe-Gln▾Gly-Pro and cleaves after the glutamine residue. The recombinant enzyme is engineered as a fusion protein with glutathione-S-transferase (GST) and polyhistidine (6xHis) tags, so that the protease can be easily removed after cleavage using a variety of solid supports with immobilized glutathione (GSH) or metal chelates, respectively. HRV3C is also active in a variety of commonly used buffers, providing further flexibility in experimental design.
Highlights:
Effective – HRV 3C is designed to remove purification tags from your protein of interest
High activity at 4°
C
– activity is ≥2000U/mg control protein; one unit will cleave 0.1mg control protein at 4°C after 16h
Flexible – dual-tagged enzyme is compatible with various columns and buffers
Includes:
Kit contains protease, cleavage buffer and control protein.
Recommended for:
Removal of purification tags from fusion proteins
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Product Line | Pierce |
| Form | Kit |
| Quantity | 1000 U |
| Product Type | Protease |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.