Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Iminobiotin Agarose PROMO

Catalog No. PI20221PM
Click to view available options
:
Thermo Scientific™ Pierce™ Iminobiotin Agarose

Agarose beads of immobilized biotin (and iminobiotin) to strongly (or reversibly) bind and remove (or purify) streptavidin or avidin proteins and conjugates from samples.

Pierce Biotin Agarose and Iminobiotin Agarose are beaded agarose supports onto which biotin and iminobiotin moieties have been covalently immobilized. Typical avidin-biotin affinity procedures use immobilized avidin or streptavidin proteins to capture biotin-tagged molecules from a sample. Occasionally, however, the reverse situation arises in which immobilized biotin in needed to capture avidin or streptavidin components from a sample. These biotin and iminobiotin resins address this special situation.

Immobilized biotin binds avidin and streptavidin very strongly, making it appropriate for non-reversible binding applications. Immobilized iminobiotin is the guanido analog of biotin. The dissociation constant of the avidin-iminobiotin complex is pH dependent. At pH 9.5 to 11, the avidin-iminobiotin complex will bind tightly. At pH 4, the avidin-iminobiotin complex will dissociate. Thus, iminobiotin agarose can be used for situations requiring reversible binding and elution of avidin components.

Highlights:

  • Enable purification or removal of avidin or streptavidin samples
  • Resins are crosslinked beaded agarose onto which biotin or iminobiotin are covalently immobilized
  • Use Biotin Agarose for non-reversible binding (e.g., removal of avidin components from a sample)
  • Use Iminobiotin Agarose for reversible binding (e.g., non-denaturing purification of avidin)

Specifications

Content And Storage Upon receipt store at 4°C.
Product Line Pierce™
Quantity 5 mL
Target Streptavidin
Type Agarose
Stationary Phase Biotin Derivative
Format Bottle
Column Type Affinity, Agarose Resin

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.