Learn More
Thermo Scientific™ Pierce™ IP Lysis Buffer, 100 mL PROMO
Description
The IP Lysis Buffer is a mammalian whole cell lysis reagent based on a modified RIPA buffer formulation without SDS. This moderate-strength lysis buffer effectively solubilizes cellular proteins but does not liberate genomic DNA or disrupt protein complexes like ordinary RIPA buffer. Pierce IP Lysis Buffer is specially formulated for pulldown and immunoprecipitation assays.
Highlights:
- Optimized for compatibility with immunoprecipitation and pull-down assays
- Compatible with protein assays, reporter assays and immunoassay procedures
- Ready-made formula is effective for extracting cytoplasmic, membrane and nuclear proteins
- Gentle formulation helps maintain protein complexes for co-immunoprecipitation (co-IP)
- Does not liberate genomic DNA which can cause high sample viscosity
Specifications
Specifications
| Quantity | 100 mL |
| Format | Liquid |
| Product Type | IP Lysis Buffer |
| Volume (Metric) | 100 mL |
| Content And Storage | Upon receipt store at 4°C. |
| Product Line | Pierce |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.