Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ IP Lysis Buffer, 250 mL PROMO
Click to view available options
:
Thermo Scientific™ Pierce™ IP Lysis Buffer
Description
The IP Lysis Buffer is a mammalian whole cell lysis reagent based on a modified RIPA buffer formulation without SDS. This moderate-strength lysis buffer effectively solubilizes cellular proteins but does not liberate genomic DNA or disrupt protein complexes like ordinary RIPA buffer. Pierce IP Lysis Buffer is specially formulated for pulldown and immunoprecipitation assays.
Highlights:
- Optimized for compatibility with immunoprecipitation and pull-down assays
- Compatible with protein assays, reporter assays and immunoassay procedures
- Ready-made formula is effective for extracting cytoplasmic, membrane and nuclear proteins
- Gentle formulation helps maintain protein complexes for co-immunoprecipitation (co-IP)
- Does not liberate genomic DNA which can cause high sample viscosity
Specifications
Specifications
Quantity | 250 mL |
Format | Liquid |
Product Type | Lysis Buffer |
Volume (Metric) | 250 mL |
Content And Storage | Upon receipt store at 4°C. |
Product Line | Pierce™ |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction