Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Jacalin Agarose PROMO

Catalog No. PI20395PM
Click to view available options
:
Thermo Scientific™ Pierce™ Jacalin Agarose

Purify human IgA from serum or colostrum or efficiently remove contaminating IgA from IgG samples with this specific affinity resin.

Jacalin is an alpha-D-galactose binding lectin extracted from jack-fruit seeds (Artocarpus integrifolia). This lectin is a glycoprotein (approx. 40kDa) composed of four identical subunits. Jacalin binds to human IgA1 (also called serum or secretory IgA) with high specificity. Thus, Jacalin that has been immobilized on crosslinked beaded agarose is useful for purifying human IgA from IgG and IgM in serum or colostrum samples.

Highlights:

  • Immobilized Jacalin – specifically binds human IgA1 (not human IgA2 or other the IgA from other species)

  • Binding capacity – resin is loaded with approx. 4.5mg Jacalin/mL resin and effectively binds 1 to 3mg human IgA1/mL resin

  • Agarose resin – support is 6% crosslinked beaded agarose, the most popular and versatile resin for protein affinity purification methods

Recommended for:

Purify human IgA1 from serum or colostrum; purify or remove IgA from immunoglobulin samples; Separate IgA1 from IgA2

Specifications

Product Type Agarose
Product Line Pierce™
Column Type Agarose Resin, Affinity
Format Bottle
Stationary Phase Lectin
Purification Target IgA Antibody
Content And Storage Upon receipt store product at 4°C. Product is shipped at ambient temperature. Do not freeze product.
Quantity 5 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.