Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Kinase Enrichment Kit with ATP Probe PROMO

Catalog No. PI88310PM
Click to view available options
:
Thermo Scientific™ Pierce™ Kinase Enrichment Kit with ATP Probe

Desthiobiotin-ATP and -ADP probes to label, affinity purify and measure the active site of ATPases (kinases), including chaperones and metabolic enzymes.

These products, based on ActivX ATP and ADP Probes, feature an amine-reactive nucleotide analog and a desthiobiotin (biotin analog) tag that facilitates selective labeling of lysines in the kinase active site and then subsequent enrichment and recovery of labeled protein. These features allow identification and profiling of target enzyme classes across samples or assessment of the specificity and affinity of enzyme inhibitors.

Highlights:

  • Specific – label only the conserved active-site lysines of nucleotide-binding proteins

  • Flexible – use for in vitro labeling of ATPase enzymes derived from cells or tissues

  • Compatible – use with Western blot or mass spectrometry (MS) workflows

Recommended for:

Profile small-molecule binding affinities and active-site inhibition in a dose-dependent manner; Identify dozens to hundreds of inhibitor targets and off-targets from tissues, cells and subcellular proteomes; Enrichment of enzymes based on function.

Specifications

Formulation Multi-component Kit with Desthiobiotin-ATP Probe
Content And Storage ActivX Desthiobiotin-ATP Probe is shipped separately with dry ice and stored at -80°C upon receipt. All other Kit components are shipped at ambient temperature and stored at 4°C upon receipt.
Assay Protein Quantitation Assay
Detection Method Chemiluminescence, Mass Spectrometry, Western Blot
Substrate Type Kinase Substrate
Product Line ActivX™, Pierce™
Quantity 16 Reactions
For Use With (Application) Pull-Down
Target Kinases (Multiple), Phosphodiesterases, ATPases, GTPases, Phosphatases
Type Kinase Enrichment Kit
Label or Dye Biotin and Other Hapten
Format Tube, Spin Column
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.