Learn More
Thermo Scientific™ Pierce™ LTQ Velos ESI Positive Ion Calibration Solution PROMO
Description
The Pierce LTQ Velos ESI Positive Ion Calibration Solution is a ready-to-use liquid formulation ideal for quickly performing the routine calibration required to maintain the robust performance of Thermo Scientific Mass Spectrometers. The LTQ Velos ESI Positive Ion Calibration Solution is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.
Highlights:
Strong peaks – mixture of caffeine, MRFA, Ultramark 1621 and n-butylamine in a acetonitrile/methanol/acetic solution
Ready-to-use – load the mass reference standard into a syringe and inject into the instrument
High-purity – mass spectrometry grade reagents in a non-leachable container
Stable – store at room temperature for more than one year
Specifications
Specifications
| For Use With (Equipment) | Mass Spectrometer |
| Product Type | Calibration Solution |
| Product Line | Pierce |
| Content And Storage | Store between 2°C and 8°C. |
| Starting Material | Peptides |
| Final Product Type | Peptides |
| Detection Method | Mass Spectrometry |
| Workflow Step | Calibration |
| Quantity | 10 mL |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.