Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Maleimide Activated Plates

Catalog No. p-7227777
Click to view available options
Color:
Black
Clear
White
Format:
Standard
Strip Plate
Detection Method:
Chemiluminescence
Colorimetric
Fluorescence

Thermo Scientific™ Pierce Maleimide Activated Plates are ideal for binding sulfhydryl-containing molecules that are difficult to coat onto polystyrene plates, such as peptides that contain a terminal cysteine.

Pierce Maleimide Activated Plates provide covalent attachment (plate capture) of sulfhydryl-containing molecules, such as cysteine-terminated peptides. These coated plates are an especially useful tool for assessing specific anti-hapten antibody titers during antibody production. Maleimide Activated Plates are available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays.

Highlights
• Spontaneous immobilization of cysteine-containin peptides and proteins with free sulfhydryls
• Pre-blocked to reduce nonspecific binding

TRUSTED_SUSTAINABILITY

Specifications

Binding Property Standard Capacity, Covalent
Format Strip Plate
Content And Storage Note: Plates are supplied pre-blocked with bovine serum albumin.

Upon receipt store plates at 4°C protected from moisture. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature.
Detection Method Colorimetric
Product Line Pierce
Color Clear
Product Type Detection Plate, Immunoassay, ELISA
Material Polystyrene
Volume (Metric) Well 100 μL
No. of Wells 96
Surface Treatment Maleimide
No. per Pack 5 Plates
Plate Blocking Blocker™ BSA Buffer
Quantity 5 plates
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.