Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Microdialysis Plate, 48 well, 1 mL, 10K MWCO, 1 Plate Set PROMO

Catalog No. PIA50470PM
Click to view available options
:
Thermo Scientific™ Pierce™ Microdialysis Plate

Catalog No. PIA50470PM

Supplier: Thermo Scientific™ A50470

Simultaneously process 1 to 96 samples with 10 to 100 μL volumes with this convenient rapid dialysis system that is automation compatible and disposable.

Each sample processed with the Pierce Microdialysis Plate is dialyzed inside a single, removable dialysis device consisting of two regenerated cellulose membranes ∼2 mm apart. The short diffusion distance and large surface area facilitate rapid dialysis, with the removal of salts and small molecules often complete in 2–4 hours. Sample is recovered using standard laboratory micropipettes. The plate seals are made of high-quality polypropylene and help protect the samples from contamination and evaporation, especially during long-term storage.

Features of Pierce Microdialysis Plate:

  • High yield—greater than 90% protein recovery
  • Rapid—dialysis completed in 2–4 hours
  • Easy to use—sample loading and retrieval can be done with a standard pipette tip
  • Flexible—detachable 8-device strips, scalable from 1 to 96 samples
  • Automation-compatible—plate format conforms to the SBS Microplate Standard

Each 48-well and 96-well Pierce Microdialysis Plate comes pre-loaded with twelve 8-dialysis device strips. Each dialysis device strip can be removed from the plate and separated into individual microdialysis devices, allowing only the number of needed devices to be used and eliminating waste. The sample chamber inside each microdialysis device is formed by two low-binding regenerated cellulose membranes that are rated to retain proteins and other macromolecules >2,000 Daltons while allowing removal of buffer salts and contaminants

Applications:

  • Protein and DNA desalting/buffer exchange
  • Removal of dyes after protein labeling
  • Enzyme activity/binding assays
  • Drug binding studies (also see Rapid Equilbrium Dialysis 'RED' devices)

Specifications

Product Type Microdialysis Plate, 48-well
MWCO 10 kDa
Membrane Cellulose Membrane, Dialysis
Format Microdialysis Plate
Capacity (Metric) 1 mL
Purification Target Buffer Exchange
Sample Volume (Metric) 1 mL
Sufficient For Dialysis of 1 to 48 samples, 150–1000 μL
Quantity 1 Plate Set
Content And Storage Product shipped at ambient temperature. Upon receipt, store at room temperature.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.