Learn More
Thermo Scientific™ Pierce™ MS-Compatible Magnetic IP Kit, Streptavidin, 40 reactions kit PROMO
Description
- MS-compatible—reagents directly compatible with in-solution peptide digestion
- Sensitive—procedure successfully enriches low-abundance proteins (low ng range)
- Low background—enriched samples contain minimal detergent residuals
- Reduced antibody contamination—antibody biotinylation with streptavidin enrichment reduces contamination in the eluate when compared to other IP methods
- Robust—procedure and reagents have been verified with numerous targets to ensure consistent enrichment of low-abundance proteins (ng range) with at least two peptides identified per protein
The Thermo Scientific Pierce MS-Compatible Magnetic IP Kit (Protein A/G) uses high-quality Pierce Protein A/G Magnetic Beads to provide wider flexibility of antibody capture than either Protein A or G alone. The Thermo Scientific Pierce MS-Compatible Magnetic IP Kit (Streptavidin) uses high-quality Pierce Streptavidin Magnetic Beads for use with biotinylated antibodies that may be purchased commercially or labeled using the Pierce Antibody Biotinylation Kit for IP. After the immunoprecipitation procedure, the target-enriched elution fraction is ready for in-solution tryptic digestion for LC-MS analysis without the need for gel purification, detergent removal, or desalting. Additionally, the reagents and procedures have been validated using both manual and automated magnetic separation.
Applications
- Enrichment of low-abundance targets to improve MS sensitivity
- Identification of target proteins that are not detectable by western blotting
- Quantitation of target proteins by LC-MS analysis
- Screening of antibodies for IP-MS
Specifications
Specifications
For Use With (Equipment) | KingFisher™ Flex, Magnetic Stand |
Product Type | Magnetic IP Kit |
Product Line | Pierce™ |
Sufficient For | 40 Reactions |
Content And Storage | Streptavidin Magnetic Beads, 1 mL, IP-MS Cell Lysis Buffer, 100 mL, IP-MS Wash Buffer A, 75 mL, IP-MS Wash Buffer B, 40 mL, IP-MS Elution Buffer, 6 mL Store in refrigerator (2° to 8°C). |
Starting Material | Protein Samples, Cell Lysate |
Final Product Type | Protein |
Detection Method | Mass Spectrometry |
Format | Kit |
Workflow Step | Sample Enrichment |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.