Learn More
Thermo Scientific™ Pierce™ Ni-NTA Magnetic Agarose Beads, 20 mL PROMO
Description
Product features:
Ni-NTA Magnetic Agarose Beads are used for small-scale affinity purification as well as high-throughput screening of recombinant His-tagged proteins. The polyhistidine tag is the most popular affinity tag and typically consists of six consecutive histidine residues (6xHis). Tagged proteins are overexpressed in a number of different systems, most commonly in bacteria, and purified from cell lysates such as those prepared using Thermo Scientific B-PER Bacterial Protein Extraction reagents. Purification of His-tagged proteins is achieved using an NTA chelate charged with nickel that coordinates with the histidine side chains. The NTA chelate contains four metal-binding sites that allow for low metal ion leaching and high binding capacity. The protocol for the Ni-NTA Magnetic Agarose Beads has been optimized to allow for high purity of the isolated His-tagged protein.
- High capacity—sufficient for both routine and demanding purification procedures; binds >70 mg of 6xHis-tagged protein per mL of settled resin
- High-performance and reproducible beads—non-aggregating, magnetite (Fe3O4), superparamagnetic beads provide exceptional uniformity for both manual and automated HTS purification applications
- Low nonspecific binding—optimized purification protocol results in: better tagged-protein purification
- Versatile—purify proteins using native or denaturing conditions
- Compatible—use with Thermo Scientific Cell Lysis reagents and a variety of buffer additives
Applications:
- Protein purification and isolation
- High throughput protein purification using automated magnetic particle processor
Target molecule: His-Tag
Specifications
Specifications
| Content And Storage | 5 mL settled resin. Supplied as 20 mL of 25% v/v suspension in 20% ethanol. Store at 2°C to 8°C | 
| Shipping Condition | Wet Ice | 
| Form | Liquid Suspension | 
| Product Line | Pierce™ | 
| Quantity | 20 mL | 
| Protein Form | Recombinant | 
| Type | Magnetic Agarose Bead | 
| Stationary Phase | Metal Chelate, Divalent Nickel | 
| Format | Bottle | 
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
