Learn More
Thermo Scientific™ Pierce™ p-Aminophenyl Phosphoryl Choline Agarose PROMO
Description
Immobilized phosphoryl choline is a beaded agarose affinity resin for purification of C-reactive protein (CRP) from plasma, ascites and other biological fluids. CRP is a pentameric (pentraxin) protein that is classified as an acute phase reactant because of its high level of production during inflammation. The protein binds to phosphoryl choline groups on the surface of microbes and has been linked to several biological functions including activation of the classical complement pathway, enhancement of phagocytosis and interaction with certain subpopulations of T-lymphocytes. In the late 1970s, Volanakis and colleagues discovered a method of isolating and studying C-reactive protein by using the protein's affinity for phosphoryl choline that has been immobilized to a porous solid chromatography support.
Highlights:
Ligand – p-aminophenyl phosphoryl choline
Support – crosslinked 6% beaded agarose
Binding capacity – greater than 3mg of human C-reactive protein per milliliter of settled resin
Format – 50% resin slurry in 0.02% sodium azide
Binding Condition – Tris or borate-buffered saline (pH 8 to 8.5) with 1 to 2mM calcium chloride
Elution Condition – Tris or borate-buffered saline (pH 8 to 8.5) with 2mM EDTA
Reusable – resin can be regenerated and reused several times
Recommended for:
Purify c-reactive protein (CRP); Purify anti-p-Aminophenyl phosphoryl choline antibodies
Specifications
Specifications
Content And Storage | Upon receipt store at 4°C. |
Form | Liquid |
Product Line | Pierce™ |
Quantity | 5 mL |
Target | C-reactive Protein |
Type | Agarose |
Stationary Phase | p-Amino Phosphoryl Choline |
Format | Bottle |
Column Type | Affinity, Agarose Resin |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.