Learn More
Thermo Scientific™ Pierce™ Peptide Retention Time Calibration Mixture PROMO
Description
The Pierce Peptide Retention Time Calibration Mixture is used to optimize and assess LC parameters, identify total peptide elution window and optimize MS parameters. Testing the mixture frequently throughout the lifetime of a column or including it in quantitative samples makes it possible to monitor and normalize LC-MS performance between samples and over time. When used with Thermo Scientific Pinpoint 1.1 software, the calibration mixture is used to predict peptide retention times based upon peptide sequence and ease the transfer of methods between LC-MS platforms.
Highlights:
- Assessment of chromatography and MS instrument performance
- Prediction of peptide retention across multiple instrument platforms
- Prediction of peptide retention time from sequence using calculated hydrophobicity factor
- Optimization of scheduled MS acquisition windows for improved quantification and increased multiplexing
- Internal standard to normalize for variation in retention times and peak intensities between runs
Specifications
Specifications
| Formulation | Mixture of 15 synthetic heavy peptides at 0.5uM each |
| Product Type | Peptide Retention Time Calibration Mixture |
| Sufficient For | 5 to 100 uses |
| Quantity | 50 μL |
| For Use With (Application) | LC-MS |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.