Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Peptide Retention Time Calibration Mixture PROMO

Catalog No. PI88321PM
Change view
Click to view available options
:
Thermo Scientific™ Pierce™ Peptide Retention Time Calibration Mixture
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PI88321PM Thermo Scientific™ Pierce™ Peptide Retention Time Calibration Mixture
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88321PM Supplier Thermo Scientific™ Supplier No. 88321

15 synthetic heavy peptides mixed in equimolar ratio to test and calibrate reverse-phase liquid chromatography (LC) and mass spectrometry (LC-MS) equipment.

The Pierce Peptide Retention Time Calibration Mixture is used to optimize and assess LC parameters, identify total peptide elution window and optimize MS parameters. Testing the mixture frequently throughout the lifetime of a column or including it in quantitative samples makes it possible to monitor and normalize LC-MS performance between samples and over time. When used with Thermo Scientific Pinpoint 1.1 software, the calibration mixture is used to predict peptide retention times based upon peptide sequence and ease the transfer of methods between LC-MS platforms.

Highlights:

  • Assessment of chromatography and MS instrument performance
  • Prediction of peptide retention across multiple instrument platforms
  • Prediction of peptide retention time from sequence using calculated hydrophobicity factor
  • Optimization of scheduled MS acquisition windows for improved quantification and increased multiplexing
  • Internal standard to normalize for variation in retention times and peak intensities between runs

Specifications

Formulation Mixture of 15 synthetic heavy peptides at 5uM each
For Use With (Equipment) Mass Spectrometer
Product Type Peptide Retention Time Calibration Mixture
Product Line Pierce
Content And Storage Upon receipt store at -80°C.
Starting Material Peptides
Final Product Type Peptides
Detection Method Mass Spectrometry
Workflow Step Calibration
Quantity 200 μL
Form Liquid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.