Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ Plus Activated Peroxidase

Catalog No. p-4532208
Click to view available options
Quantity:
1 mg
5 x 1mg

Periodate-aldehyde-activated horseradish peroxidase (HRP) enzyme and kit to label primary antibodies to obtain high-activity antibody-HRP conjugates.

Thermo Scientific™ EZ-Link™ Plus Activated Peroxidase Kit contains horseradish peroxidase enzyme that has been aldehyde-activated with periodate to enable high-efficiency labeling and preparation of protein or antibody conjugates.

Other amine-reactive chemistries, like glutaraldehyde, have a tendency to polymerize and produce lower amounts of viable conjugate. Sugar residues present on HRP have been oxidized with periodate to produce aldehydes that react with primary amines. Plus Activated Peroxidase is mixed with the protein to be coupled at a pH compatible with your protein or antibody. After incubating the reaction for one to two hours, the conjugate linkage is reduced and the activated HRP quenched with ethanolamine. The conjugate is purified by desalting or by dialysis. If you are preparing an antibody-HRP conjugate, use the Thermo Scientific Pierce Conjugate Purification Kit (Part No. 44920) to separate unreacted enzyme from the conjugate.

Highlights:

  • Reacts with readily accessible primary amines to form a covalent secondary amine bond
  • Can be stored for at least 12 months at -20°C
  • Consistent conjugation yields reliable conjugates
  • Enzyme activity is 120-200 units/mg
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine
Quantity 1 mg
Solubility Water
Reactive Moiety Aldehyde
Content And Storage Note: EZ-Link Plus Activated Peroxidase contains 1-3 moles of aldehyde per mole of peroxidase.

Storage: Upon receipt store product protected from moisture at -20°C. Product is shipped with an ice pack.
Form Solid
Product Line EZ-Link™
Label Type Enzyme Labeled
Label or Dye HRP (Horseradish Peroxidase)
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.