Learn More
Thermo Scientific™ Pierce™ Prestained Protein MW Marker

Description
Thermo Scientific™ Prestained Protein Molecular Weight Marker is a mixture of six blue-stained natural proteins (20 to 120kDa) for use as size standards in protein electrophoresis (SDS-PAGE).
Thermo Scientific Prestained Protein Molecular Weight Marker is a mixture of six blue-stained natural proteins (20 to 120 kDa) for use as size standards in protein electrophoresis (SDS-PAGE). The protein standards are supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.
Component proteins
- Beta-galactosidase (120 kDa)
- Bovine serum albumin (85 kDa)
- Ovalbumin (50 kDa)
- Cardonic anhydrase (35 kDa)
- Beta-lactoglobulin (25 kDa)
- Lysozyme (20 kDa)
Applications
- Monitoring protein migration during SDS-polyacrylamide gel electrophoresis.
- Monitoring protein transfer onto membranes after western blotting
- Sizing of proteins on SDS-polyacrylamide gels and western blots

Specifications
Specifications
Content And Storage | Storage Buffer: 62.5 mM Tris · H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% (w/v) SDS, 1 mM DTT, 1.5 mM NaN3 and 33% (v/v) glycerol. Upon receipt store at -20°C. Product is shipped with an ice pack. |
Detection Method | Colorimetric |
Number of Markers | 6 |
Product Type | Protein Molecular Weight (MW) Marker |
Ready to Load | Yes |
Size Range | 20 to 120 kDa |
Stain Type | 1 color: Blue |
Molecular Weight (g/mol) | 120, 85, 50, 35, 25, 20 kDa |
Product Line | Pierce™ |
Quantity | 8 x 250 μL |
Show More |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.