Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Protein A/G IgG Binding Buffer PROMO

Catalog No. PI54200PM
Click to view available options
:
Thermo Scientific™ Pierce™ Protein A/G IgG Binding Buffer

Catalog No. PI54200PM

Supplier: Thermo Scientific™ 54200

Mouse Monoclonal Secondary Antibody

These products include IgG Binding Buffers specific for each immunoglobulin-binding protein (Protein A, Protein G, Protein A/G). The Pierce IgG Elution Buffer provides efficient dissociation and elution of antibodies from all three IgG-binding proteins, including Protein L. This low-pH elution buffer is also effective for most antibody-antigen or protein-protein affinity purification systems.

The Gentle Ag/Ab Binding and Elution Buffer system uses near-neutral, high-salt conditions for elution, enabling even sensitive and labile antibodies (or other proteins) to be eluted from an affinity system without denaturation and inactivation. The phosphate-free binding buffer in this system is a necessary substitute for the Protein A binding buffer (pH 8), which precipitates with the Gentle Ag/Ab Elution Buffer.

Finally, the Mouse IgG1 Mild Elution Buffer, when used in combination with Protein A resin, enables selective elution and separation of the IgG1 subclass from total bound mouse IgG.

Highlights:

  • Three optimized binding buffers – for Protein A, Protein G or Protein A/G

  • Two elution buffer options – acidic (pH 2.8) or high salt (near-neutral)

  • Special system for mouse IgG1 – selective, two-stage elution from Protein A agarose

Item-Specific Features:

  • Protein A IgG Binding Buffer: pH 8.0; phosphate-based; contains glycerol and EDTA

  • Protein A/G IgG Binding Buffer: pH 8.0; phosphate-based; contains EDTA

  • Protein G IgG Binding Buffer: pH 5.0; contains 0.02% sodium azide

  • IgG Elution Buffer: pH 2.8; amine-based

  • Gentle Ag/Ab Binding Buffer: pH 8.0; phosphate-free; contains 0.05% sodium azide

  • Gentle Ag/Ab Elution Buffer: pH 6.6; high salt (ionic strength)

  • Mouse IgG1 Mild Elution Buffer: pH 6.0; contains 0.02% sodium azide

Includes:

Kits contain both binding and elution buffers

Specifications

Chemical Name or Material Binding Buffer
pH 8
Physical Form Liquid
Product Line Pierce
Quantity 240 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.