Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Protein A/G Magnetic Beads, 1 mL PROMO

Catalog No. PI88802PM
Click to view available options
:
Thermo Scientific™ Pierce™ Protein A/G Magnetic Beads

Immunoprecipitate or affinity purify antibodies (IgG) using Protein A/G coated magnetic particles for manual or robotic magnetic separation.

These magnetic beads are coated with genetically engineered Pierce Protein A/G, a recombinant fusion protein which combines the IgG binding domains of both Protein A and Protein G. This enables capture of antibodies from a wider range of species and isotypes than either protein alone. Using our crosslinker chemistry, you can immobilize an antibody onto the magnetic particle and prevent IgG contamination in your immunoprecipitated sample. These beads can be used both manually with a magnetic stand as well as with automated platforms such as the Thermo Scientific KingFisher™ Instruments.

Highlights:

  • High capacity – nearly four times higher binding capacity than typical magnetic beads from other suppliers, allowing the use of smaller amounts per experiment

  • Low non-specific binding – stable, pre-blocked beads provide clean purification products (e.g., antigen eluted in IP with antibody is devoid of contaminating proteins from complex IP matrix)

  • Flexibility – convenience of IgG binding domains of both Protein A and Protein G on one bead

  • Compatibility - beads are compatible with manual and automated applications (e.g., Thermo Scientific KingFisher Instruments)

  • Assay consistency – magnetic beads eliminate resin loss and provide for more efficient separation of solutions than traditional IP methods that use only microcentrifuge tubes

Properties of Thermo Scientific Pierce Protein A/G Magnetic Beads:

  • Composition: Magnetite-coated polymeric beads blocked and covalently coated with a monolayer of recombinant Protein A/G
  • Diameter: 1μm
  • Concentration: 10 mg/mL
  • Binding Capacity: 55-85μg rabbit IgG per mg magnetic particles

Requires:

Thermo Scientific KingFisher 96 or KingFisher Flex Instrument or other magnetic separator device

Recommended for:

IP and Co-IP experiments (see complete kit); Immunoprecipitation for analysis in non-reducing conditions; Antibody purification

Specifications

Content And Storage Upon receipt store at 4°C. Product shipped with an ice pack.
Form Liquid
Product Line Pierce™
Quantity 1 mL
Protein Form Recombinant
Target Antibody
Type Magnetic Affinity Bead
Ligand Type Protein A/G
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.