Learn More
Thermo Scientific™ Pierce™ Protein Concentrator PES, 10K MWCO, 5-20 mL, 24 Concentrators PROMO
Description
For protein and other biological samples, such as nucleic acids, with a 10K MWCO, Pierce Protein Concentrators PES offer rapid protein concentration, excellent sample recovery, and instrument compatibility. These centrifugal devices feature a high-flux polyethersulfone (PES) membrane for concentration, desalting, and buffer exchange of biological samples. These concentrators have a MWCO of 10K and can be used for processing sample volumes from 0.5 to 100 mL.
Features of 10K MWCO Protein Concentrators PES include:
- Rapid protein concentration—sample concentration of 10- to 30-fold can be achieved in 10 or fewer minutes using the 0.5-mL concentrator and 15 or fewer minutes using the other quantity concentrators
- Excellent protein recovery—typical protein recovery is >90% (when the sample mass is at least two-fold greater than the posted MWCO)
- Instrument-compatibility—a standard microcentrifuge with swinging (except 88513) or fixed-angle rotors can be used; sample can be collected and recovered without invert spinning
- Versatile—easily perform sample concentration, diafiltration, or buffer-exchange of biological samples
For easy identification, the MWCO value is etched on the side of each device. To assist with sample addition and recovery, a clear window with graduations is located on the side of each device, enabling visual determination of the retentate volume. The vertical design of the membrane allows high-flux rates, low non-specific binding, and negligible protein aggregation on the membrane, giving reliable and consistent results for processing sample volumes between 0.5 and 100 mL. Typical protein recovery is 90% when the mass of the proteins in the sample is at least two-fold greater than the MWCO of the device used. For example, when concentrating IgG (MW ∼150 KDa) samples, use the 3K, 10K, 30K, or 50K MWCO concentrators. Final sample concentrations of 10- to 30-fold can be achieved within 10 minutes when using the 0.5 mL concentrators and within 15 minutes when using the other concentrator sizes.
Protein Concentrators PES applications
- Protein concentration of tissue culture media, antiserum, or monoclonal antibody preparations
- Concentration of protein peaks following gel-permeation chromatography
- Removal of unincorporated label following protein modification reactions
- Concentration and desalt/buffer-exchange after elution of protein from ion-exchange
- Separation of different sized molecules
Pierce Protein Concentrators PES centrifugal devices contain a membrane rated for retaining molecules with a molecular weight at least two-fold greater than the MWCO rating of the PES membrane within the device. Reduced recovery may occur when using a concentrator with molecules smaller than the recommended mass. Protein recovery will vary depending on the specific protein in the sample and its starting concentration.
The dead-stop volume for the 0.5 mL, 2–6 mL, 5–20 mL, and 20–100 mL concentrators is approximately 15 μL, 30 μL, 50 μL, and 350 μL, respectively.
To achieve >90% recovery of protein with the 0.5 mL concentrators, the minimum protein sample concentration should be 0.05 mg/mL. For all other concentrators, the minimum protein sample concentration should be 0.1 mg/mL.
Optimal centrifuges and speeds
The 0.5-mL concentrators can be used effectively at a maximum centrifugal force of 15,000 x g, using a fixed angle centrifuge only.
For the 2-6 mL concentrators, swinging-bucket or fixed-angle centrifuge models can be used. Swinging bucket centrifuges capable of generating 2,000–4,000 x g and a maximum centrifugal force of 4,000 x g are recommended. With fixed-angle centrifuges, models capable of generating 5,000–10,000 x g and a maximum centrifugal force of 8,000 x g are recommended.
For the 5-20 mL volume concentrators, swinging-bucket or fixed-angle centrifuge models can be used. Swinging-bucket centrifuges capable of generating 2,000–5,000 x g and a maximum centrifugal force of 4,000 x g are recommended. With fixed-angle centrifuges, models capable of generating 5,000–8,000 x g and a maximum centrifugal force of 6,000 x g are recommended.
For the 20-100 mL concentrators, only swinging-bucket centrifuges capable of generating 1,000–2,000 x g are recommended. Use a maximum centrifugal force of 2,000 x g for the 20–100 mL concentrators. The concentrators are not recommended for use in fixed-angle rotors.
Centrifugal force, temperature and sample volume, concentration and viscosity will affect filtration rate and require optimization of time for each application.
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Disposable | Single-use |
| Material (Membrane) | PES |
| MWCO | 10 kDa |
| Sample Volume | 5 to 20 mL |
| Product Line | Pierce™ |
| Purification Target | Protein |
| Quantity | 24 Concentrators |
| Shipping Condition | Room Temperature |
| Type | Filter |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.