Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Protein Concentrator PES

Catalog No. p-6806176
Click to view available options
MWCO:
10 kDa
100 kDa
5 kDa

Thermo Scientific™ Pierce™ Protein Concentrators PES are easy-to-use centrifugal devices that provide fast processing and excellent recovery of protein samples.

The Thermo Scientific™ Pierce™ concentrators are easy-to-use centrifugal devices that provide fast processing and excellent recovery of protein samples. These disposable ultrafiltration devices contain a polyethersulfone (PES) membrane for the concentration, desalting, and buffer exchange of biological samples, such as tissue culture media, antiserum, monoclonal antibody preparations, and chromatography fractions. They can also be used to remove unincorporated label following protein modification reactions. 

The PES membrane in these devices is available in five distinct molecular-weight cutoffs (MWCO) of 3K, 5K, 10K, 30K, and 100K, and can be used for processing sample volumes from 100 μL to 100 mL. The MWCOs are etched on the sides of the concentrators for easy identification, while a clear window with graduations on the side of each device allows for visual determination of the concentrated sample (retentate) volume. The unique design provides reliable and consistent results. Multiple device sizes are available to handle maximum sample volumes of 0.5 mL, 6 mL, 20 mL, and 100 mL.
  • Rapid processing—unique design minimizes membrane fouling and sample concentration of 10- to 30-fold can be achieved in 5–30 minutes for 10K MWCO (device dependent—times may vary for other MWCO’s), even with particle laden solutions.
  • High recovery—retain >90% of protein samples while removing contaminants or exchanging buffers.
  • Convenient—clear markings, wide sample chamber and removable filtrate chamber make handling simple and easy.
  • Instrument compatible—can be used with standard centrifuges utilizing either fixed-angle or swinging-bucket rotors.
TRUSTED_SUSTAINABILITY

Specifications

Material (Membrane) PES
MWCO 5 kDa
For Use With (Application) concentration, desalting, and buffer exchange of biological samples
Quantity 1pk
Type Spin Column, Bottle
Volume (Metric) 20 to 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.