Learn More
Thermo Scientific™ Pierce™ Protein-Free Blocking Buffer
Description
Pierce Protein-Free Blocking Buffer is a non-protein compound that provides effective blocking for membrane-based and plate-based protein detection methods, resulting in extremely low background. It is supplied in a convenient, ready-to-use format.
Features of Protein-Free Blocking Buffer include:
- Protein-free blocker—minimizes or eliminates cross-reactivity associated with protein-based blocking buffers
- Applications—compatible with western blotting (membranes), ELISA (microplates), and arrays (coated glass slides)
- Streptavidin-friendly—free of biotin; no interference with avidin-biotin detection systems
- High-performance—optimized and validated in many protein methods to provide high signal-to-noise ratio (i.e., no quenching of specific binding and signal but eliminating nonspecific binding and background)
Traditional blocking buffers contain proteins that can cross-react in certain immunodetection systems, resulting in high background and reduced signal. Pierce Protein-Free Blocking Buffers are devoid of protein while remaining highly effective at blocking plates, membranes, and other surfaces for ELISA, western blotting, glass slide arrays, and other applications.
Specifications
Specifications
| Chemical Name or Material | Protein-Free T20 (TBS) |
| Concentration | 1X |
| For Use With (Application) | ELISA |
| Physical Form | Liquid |
| Product Line | Pierce |
| Quantity | 1 L |
Frequently Asked Questions (FAQs)
Optimize the concentration of primary and secondary antibodies. In some cases, increasing the concentration of blocking agent (BSA or non-fat dry milk) or usiing an alternative blocking solution such as Starting Block or SuperBlock may reduce background signal. After incubation with the primary antibody, wash at least 2 times with TBST (include 0.5 M NaCl in one or more of the wash steps). Avoid Nonidet P40 or Triton X-100 in buffers because protein detection is decreased when these detergents are used.
Yes, it is animal-origin free (AOF).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.