Learn More
Thermo Scientific™ Pierce™ Protein G Magnetic Beads, 5 mL PROMO
Description
Recombinant protein G, covalently attached to a blocked magnetic bead surface, can bind to antibodies from many different species, enabling purification of antibodies from crude extracts. Immunoprecipitation assays performed with protein G coated beads result in high yield of target antigen with very low background. Using our crosslinker chemistry, you can immobilize an antibody onto the magnetic particle and prevent IgG contamination in IP or Co–IP experiments. The Pierce Protein G Magnetic Beads can be used both manually with a magnetic stand as well as with automated platforms such as the Thermo Scientific KingFisher Instruments for high-throughput workflows.
Highlights:
High efficiency – equivalent or higher yield of IP target antigens than magnetic beads from other suppliers
Low non-specific binding – stable, pre–blocked beads provide highly purified product (e.g., antigen eluted in IP with antibody is devoid of contaminating protein from complex cell lysate)
Consistent – magnetic beads eliminate resin loss and provide for more efficient separation of solutions than traditional IP methods that use only centrifugation
Versatile – beads are compatible with manual and automated workflows (e.g., Thermo Scientific™ KingFisher Instruments)
Recommended for:
- IP and Co-IP experiments
- Antibody purification
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. Product shipped with an ice pack. |
| Form | Liquid |
| Product Line | Pierce |
| Quantity | 5 mL |
| Protein Form | Recombinant |
| Target | Antibody |
| Type | Magnetic Affinity Bead |
| Ligand Type | Protein G |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.