Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Protein G Magnetic Beads
GSA_VA

Catalog No. p-4589197
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88848 5 mL
PI88847 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI88848 Supplier Thermo Scientific™ Supplier No. 88848
Only null left
Add to Cart
Edge
Add to Cart

Perform antibody purification and immunoprecipitation methods using manual or robotic magnetic separators with these high-capacity and high-throughput affinity particles.

Thermo Scientific Pierce Protein G Magnetic Beads are high-capacity and high-throughput affinity particles for antibody purification and immunoprecipitation methods using manual or robotic magnetic separators.

Features of Protein G Magnetic Beads:

  • High efficiency – equivalent or higher yield of IP target antigens than magnetic beads from other suppliers
  • Low non-specific binding – stable, pre-blocked beads provide highly purified product (e.g., antigen eluted in IP with antibody is devoid of contaminating protein from complex cell lysate)
  • Consistent – magnetic beads eliminate resin loss and provide for more efficient separation of solutions than traditional IP methods that use only centrifugation
  • Versatile – beads are compatible with manual and automated workflows (e.g., Thermo Scientific KingFisher Instruments)

Recombinant protein G, covalently attached to a blocked magnetic bead surface, can bind to antibodies from many different species, enabling purification of antibodies from crude extracts. Immunoprecipitation assays performed with protein G coated beads result in high yield of target antigen with very low background. Using our crosslinker chemistry, you can immobilize an antibody onto the magnetic particle and prevent IgG contamination in IP or Co-IP experiments. The Pierce Protein G Magnetic Beads can be used both manually with a magnetic stand as well as with automated platforms such as the Thermo Scientific KingFisher Instruments for high-throughput workflows.

Applications:

  • IP and Co-IP experiments
  • Antibody purification

Thermo Scientific Pierce Protein G Magnetic Beads are used for purifying antibody from serum, cell culture supernatant or ascites as well as for IP/Co-IP of antigens from cell or tissue extracts. Protein G can bind to antibodies from many different species, including mouse, human, rabbit, cow, goat and sheep. The protocol for the Pierce Protein G beads has been optimized to allow for high recovery and high purity of the isolated antibody or antigen. Antibody or antigen/antibody complex (IP) is first captured on the magnetic beads. The beads are washed and then the target is eluted with low pH elution buffer. IP performance is equivalent to or better than Protein G magnetic beads from other suppliers.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 5 mL
Concentration Slurry: 10mg/mL, 1% solids
Content And Storage Upon receipt store at 4°C. Product shipped with an ice pack.
Particle Size 1 μm
Form Liquid
Product Line Pierce
Quantity 5 mL
Protein Form Recombinant
Target Antibody
Type Magnetic Affinity Bead
Ligand Type Protein G
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.