Learn More
Thermo Scientific™ Pierce™ Protein Precipitation Plates, 1 Kit (10 Plates) PROMO
Description
Pierce™ Protein Precipitation Plates are designed specifically for solvent-based precipitation of proteins and their subsequent removal from fluid samples. The plates are leak-free and retain organic solvent without dripping, even while mixing. The 2 mL well volume allows processing sample volumes of 15 to 400 μL per well, using the recommended precipitation conditions of 3:1 (v/v) solvent-to-serum ratio. The Protein Precipitation Plates are the ideal choice for downstream processing of plasma protein binding assay samples prepared with rapid equilibrium dialysis (RED) systems that are important for the pharmaceutical industry to perform plasma protein-binding assays, which comprise a critical step in drug development. Pierce Protein Precipitation Plates use a special graded filter system that retains precipitated protein particles without clogging, yet freely allows soluble (nonprecipitated) proteins to elute through the 0.2 μM cut-off membrane. This design maximizes protein removal and ensures the highest possible level of sample recovery.
Features of Protein Precipitation Plates:
- Prepare small molecule samples for HPLC or LC-MS in 10–30 minutes
- Compatible with most organic solvents
- No acetonitrile leakage for up to four hours
- Leach-free graded hydrophobic frit prevents blockage
- Filtrates are easily collected by vacuum, positive pressure or centrifugation
- Assay serum samples of 15 to 400 μL per well (2 mL well volume)
Specifications
Specifications
For Use With (Equipment) | Mass Spectrometer, Centrifuge |
Product Type | Protein Precipitation Plate |
Product Line | Pierce™ |
Content And Storage | Upon receipt store plates at room temperature. Product shipped at ambient temperature. |
Starting Material | Protease-digested Protein |
Final Product Type | Peptides |
Detection Method | Mass Spectrometry |
Format | Plate |
Workflow Step | Sample Clean Up |
Quantity | 1 Kit (10 Plates) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.