Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Pyrogen-Free Plate Strips with Frame, 12 x 8 well PROMO

Catalog No. PIA43888PM
Click to view available options
:
Thermo Scientific™ Pierce™ Pyrogen-Free Plate Strips with Frame

The endotoxin-free 12 x 8-well plates are supplied with one frame and contain <0.0005 EU/well endotoxin. The individually wrapped 8-well strips are ideal for assays of less than 96 tests at a time.

These endotoxin-free 12 x 8-well plates are supplied with one frame and contain <0.0005 EU/well endotoxin. The individually wrapped 8-well strips are ideal for assays of less than 96 tests at a time.

Specifications

Format Strip Plate
Sterility Sterile
For Use With (Application) Chromogenic LAL Endotoxin Assay, Kinetic Chromogenic LAL Assay and Kinetic Turbidimetric LAL Assay
Quantity 5 plates, 1 frame

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.