Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IFN-alpha 2 (alpha 2b) Recombinant Protein

Catalog No. pi111051
Click to view available options
Quantity:
1 x 10^5 units

Recombinant Protein

After receipt, the product may be stored at -20°C for short-term use (less than 6 months). For long-term storage, store the product at -70°C or below for retention of full activity. Thaw product vial by incubation in cold tap water until just thawed. Aliquot and refreeze unused contents to minimize effects of freeze thawing. Refreezing should be done on dry ice or in a dry ice/alcohol bath. Further dilution of product should be in buffers containing protein such as 0.1% BSA or tissue culture media with serum.

Interferon alpha-2b is a cytokine that belongs to the type I interferon family and is encoded by a gene cluster on chromosome 9. It is produced in response to viral infection and plays a key role in the innate immune response, exhibiting potent antiviral, antiproliferative and immunomodulatory properties. Interferon alpha-2b binds to a plasma membrane receptor consisting of IFNAR1 and IFNAR2, which is expressed ubiquitously in the body and can thus act on virtually all cells. This cytokine is effective in reducing the symptoms and duration of the common cold, and has shown efficacy in treating various types of cancer, including hematological malignancies and solid tumors. A deficiency of type I interferon in the blood is considered a hallmark of severe COVID-19, and a combination therapy involving interferon alpha-2b may be a potential therapeutic approach. IFNA2 has been associated with diseases such as Severe Covid-19 and Hepatitis B.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01563
Concentration Lot-specific
For Use With (Application) ELISA
Formulation PBS with 0.1% BSA and no preservative
Gene ID (Entrez) 3440
Molecular Weight (g/mol) 19.3 kDa
Name Human IFN-alpha 2 (alpha 2 b)
Quantity 1 x 10^5 units
Source E. Coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias alpha-2 A interferon; IFNA; IFNA2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha-2; IFN-alphaA; INFA2; Interferon; interferon alpha 2; interferon alpha 2 A; interferon alpha 2 b; interferon alpha A; interferon alpha-2; Interferon alpha-A; interferon, alpha 2; leIF A
Common Name Interferon alpha-2 b
Gene Symbol IFNA2
Biological Activity EC50 ̃ 1 U/mL; determined by use of the cytopathic effect inhibition assay described in [Rubinstein, S. et al. (1981). J. Virol. 37, 755-758 ]; [Familletti, P.C. et al. (1981). Methods in Enzymology, Vol. 78, 387-394 ]. The units are determined with respect to the international reference standard for human interferon alpha A (Hu-IFN-alpha-A) provided by the National Institutes of Health [see Pestka, S. (1986). Methods in Enzymology, Academic Press, New York 119, 14-23 ]. Unit of activity measured on bovine MDBK cells with vesicular stomatitis virus (VSV).
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.