Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Recombinant Protein L

Catalog No. PI77680
Change view
Click to view available options
Quantity:
1 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI77680 500 mg
PI21189 1 mg
PI77679 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI77680 Supplier Thermo Scientific™ Supplier No. 77680
Add to Cart
Add to Cart

Detect mouse and human antibodies containing kappa light chains in immunoassays using this purified, biotinylated or peroxidase-conjugated, recombinant Protein L.

Purified and conjugated forms of Thermo Scientific™ Pierce Recombinant Protein L enable probing and detection of mouse and human antibodies containing kappa light chains in Western blotting, ELISA, IHC and other immunoassay protocols.

Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen-binding site. This gives Protein L the ability to bind a wider range of Ig classes and subclasses than other antibody-binding proteins. Protein L will bind to all classes of Ig (IgG, IgM, IgA, IgE and IgD). Protein L will also bind single chain variable fragments (scFv) and Fab fragments. Protein L binds kappa I, III, and IV in human and kappa I on mouse.

Highlights:

  • Contains four immunoglobulin-binding domains per protein
  • Binds members of all classes of Ig (IgG, IgM, IgA, IgE and IgD) that contain kappa light chains
  • Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains
TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 mg
Product Line Pierce
Form Powder
Protein Subtype Protein L
Target Antibodies
Content And Storage Upon receipt store at 4°C.

Note: Store reconstituted product for up to one month at 4°C. For long-term storage, aliquot, add an equal volume of glycerol and store at -20°C.
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.