Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Renilla Luciferase Flash Assay Kit

Catalog No. p-4532168
Click to view available options
Quantity:
100 Reaction Kit
1000 Reaction Kit

Assay regulatory element transcriptional activity in whole cell lysates with this highly sensitive flash-based Renilla luciferase kit.

The Thermo Scientific™ Pierce Renilla Luciferase Flash Assay Kit provides researchers with a highly sensitive intracellular assay for transcriptional activity of regulatory elements in mammalian whole cell lysate.

This Flash Assay Kit contains reagents for measuring the activity of Renilla luciferase in mammalian cell lysates. When used with Thermo Scientific Green Renilla Luc Plasmids, the kit provides an extremely sensitive bioluminescent reporter assay system for intracellular detection of promoter or pathway activity. The signal produced by Green Renilla Luc is considerably greater than signal from either native firefly or Renilla luciferases assayed under similar conditions. Light output captured using a luminometer can be correlated with the amount of Renilla luciferase protein produced and used to determine the activity of the promoter driving Renilla expression.

Highlights:

  • Sensitive – greater sensitivity allows utilization of smaller numbers of cells

  • Cost effective – highest sensitivity assays result in decreased reagent consumption

  • Time-saving – assays require minimal sample handling

  • Automation-friendly – amenable to high throughput screens

  • Convenient – contains a universal cell lysis buffer and optimized flash assay reagent

  • Safe – allows one to perform non-radioactive assays

Includes:

Cell lysis buffer, reaction buffer and substrate

Requires:

Renilla luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers

Recommended for:

Promoter studies for analyzing cis- regulatory elements and trans-acting factors; Drug screening; siRNA and miRNA screening; Multiplexed assays to study off-target effects; Protein localization reporter assays; Signal transduction pathway analysis; RNA splicing studies

TRUSTED_SUSTAINABILITY

Specifications

Type Assay Kit
Target Luciferase, Green Renilla Luciferase
Quantity 100 Reaction Kit
Content And Storage • Renilla Flash Assay Buffer, 5 mL, store at 4°C
• Coelenterazine (100X), 50 μL, store at -80°C
• 2X Cell Lysis Buffer, 6 mL, store at room temperature
Assay Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells Mammalian Cells
Detection Method Bioluminescence
For Use With (Equipment) Luminometer (Microplate)
Format 384-well plate, 96-well plate
Label Type Enzyme Labeled
Substrate Properties Chemical Substrate
Technique Enhanced Chemiluminescence
Substrate Type Luciferase Substrate
Product Line Pierce™
Substrate Coelenterazine
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.