Learn More
Thermo Scientific™ Pierce™ Reserpine Standard for LC-MS PROMO
Description
The Pierce Reserpine Standard for LC-MS is a pre-diluted liquid formulation ideal to quickly perform installation specifications of Thermo Scientific (and other) mass spectrometers. The standard is provided in a concentration of 100pg/μL in 50% isopropyl alcohol and requires minimal additional dilutions. The product is provided in a pack of 5 x 1 mL glass amber vials with PTFE-lined screw caps. The Pierce Reserpine Standard for LC-MS is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications.
Highlights:
Convenient – provided pre-diluted and requires minimal additional dilutions to reach target concentration for injection
Safe handling – unlike glass ampules, cap can be easily removed and replaced when withdrawing solution
High-purity – mass spectrometry grade reagent in non-leachable screw-cap vials
Stable – store at 4°C for up to one year
Specifications
Specifications
For Use With (Equipment) | Mass Spectrometer |
Product Type | Reserpine Standard |
Product Line | Pierce™ |
Content And Storage | Store in a cool, dry, well-ventilated area, protected from direct sunlight. |
Starting Material | Peptides, Small Molecules |
Final Product Type | Peptides, Small Molecules |
Detection Method | Mass Spectrometry |
Workflow Step | Calibration |
Quantity | 5 x 1 mL |
Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.