Learn More
Thermo Scientific™ Pierce™ RNA 3' End Desthiobiotinylation Kit PROMO
Description
The Pierce RNA 3'-End Desthiobiotinylation Kit is optimized for labeling the 3'-end of single-stranded RNA using T4 RNA ligase. Once labeled, the RNA can be used as a probe or target for gel-shift EMSA reactions, Northern blots and protein-RNA interaction experiments. The desthiobiotinylated RNA also can be used to enrich for RNA binding proteins (RBP) using streptavidin affinity resin, because the desthiobiotin tag binds to streptavidin in a manner which allows gentle elution of the ribonucleoprotein complex. The labeling kit is also included as a component of the Thermo Scientific Pierce Magnetic RNA-Protein Pull-Down Kit.
Highlights:
Non-radioactive
– a single biotin labels has similar detection sensitivity as radioactivity
Easy to use – RNA ligase and optimized reaction buffer are included
Economical – only a fraction of the cost purchasing synthetic biotinylated RNA probes
End-labeled – results in minimal disturbance of RNA secondary structure
Flexible
– desthiobiotin label may be used for detection or as an affinity handle for streptavidin resin
Includes:
- Desthiobiotinylated cytidine bisphosphate, T4 RNA ligase and buffer, unlabeled RNA oligonucleotide for use as a positive control, a biotinylated RNA probe standard, RNase inhibitor, glycogen, and ligation enhancing reagents
Recommended for:
- Northern blotting
- EMSA
- Enrichment of RNA binding proteins (RBP)
Specifications
Specifications
Content And Storage | Sufficient For: 20 desthiobiotinylation Reactions using 50 pmol of RNA each, Desthiobiotinylated Cytidine Bisphosphate, 40 μL, T4 RNA Ligase, 40 μL, T4 RNA Ligase Reaction buffer (10X), 100 μL, Non-labeled RNA Control, 100 μL, Biotinylated IRE RNA Control, 35 μL, RNase Inhibitor, 2 x 10 μL, DMSO, 200 μL, PEG (30%), 300 μL, Glycogen, 20 μL, Nuclease-free Water, 1.5 mL Upon receipt, Store at -20°C. |
Format | Kit |
Isolation Technology | Magnetic Bead |
No. of Reactions | 20 Reactions |
Product Line | Pierce™ |
Product Type | RNA Labeling Kit |
Quantity | 20 Reactions |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.