Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sodium Dodecyl Sulfate (SDS), Lauryl

Catalog No. p-4530934
Click to view available options
Quantity:
100 g
1 kg

Perform routine protein electrophoresis and cell lysis methods using this popular, standard-grade SDS detergent.

Thermo Scientific™ Sodium Dodecyl Sulfate (Lauryl) is standard-grade SDS detergent for use in protein polyacrylamide gel electrophoresis (PAGE).

Additional Information and Online Ordering

This lauryl-grade sodium dodecyl sulfate (SDS) is a popular anionic detergent for routine protein electrophoresis and cell lysis methods. The formulation is a mixture of several different alkyl sulfate chain lengths (C10 to C18).

Highlights:

  • Popular anionic detergent for a variety of protein methods
  • Especially useful for denaturing polyacrylamide gel electrophoresis (SDS-PAGE)
  • Common component of cell lysis buffers

Detergent properties (pure C12):

  • Molecular Weight: 288.5
  • Detergent Class: Ionic (anionic)
  • Aggregation Number: 62
  • Micelle Molecular Weight: 18,000
  • Critical Micelle Concentration (CMC): 6 to 8mM (0.1728 to 0.2304%, w/v)
  • Cloud Point: greater than 100°C
  • Dialyzable: No

Storage Requirements

  • Store in original container protected from direct sunlight in a dry, cool and well-ventilated area
  • Should be stored between the following temperatures: 20°C to 25°C
TRUSTED_SUSTAINABILITY

Specifications

Color White
Product Type Detergent
Solubility 10% (aq, w/v) solution must be clear, colorless to slightly yellow
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.
Form Powder
Reagent Type Detergent (Pure)
Quantity 100 g
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.