Learn More
Thermo Scientific™ Pierce™ Spin Columns - Snap Cap PROMO
Description
These microcentrifuge spin cups and spin columns are convenient tools for manipulating small volumes of affinity supports (5 to 500μL) for immunoprecipitation (IP) and pull-down purification assays. Five styles of polypropylene columns and matching collection tubes offer a variety of options to suit particular types of experimental needs. Add the affinity resin and sample to one of the columns and use a microcentrifuge to efficiently wash away contaminants and elute your purified sample without losing any resin in the process. Spin columns allow you to affinity-purify more protein in less time.
Highlights:
- Efficient washing of samples means fewer washes are needed to remove contaminating proteins
- Efficient elution of samples means better recovery of immunoprecipitation and assay products
- Filter cups and columns eliminate contamination of eluted samples with affinity resin
- Filter cups and columns eliminate the need to decant supernatant from pelleted debris or resin
- Spin protocols drastically reduce the time required for small-scale affinity protocols
- Low protein-binding polypropylene column construction minimizes nonspecific binding
Spin Cups-Paper Filter with Collection Tubes (Part No. 69700):
- Column volume: 600μL
- Resin volume: 20 to 300μL
- Filter type: paper, 10μm pore size
- Cap type: collection tube cap fits onto inserted spin cup
Spin Cups-Cellulose Acetate Filter with Collection Tubes (Part No. 69702):
- Column volume: 800μL
- Resin volume: 20 to 400μL
- Filter type: cellulose acetate, 0.45μm pore size
- Cap type: collection tube cap fits onto inserted spin cup
- Spin ColumnsScrew Cap with Luer-Lok
Spin Columns-Screw Cap (Part No. 69705):
- Column Volume: 900μL
- Resin volume: 20 to 400μL
- Filter type: polyethylene filter, 10μm pore size; small and large frit options for different sample sizes
- Cap type: O-ring screw top caps; press-in bottom plugs
- Luer-Lok Adapters enable use with syringe-based kits
Spin Columns-Snap Cap with Collection Tubes (Part No. 69725):
- Column volume: 1000μL
- Resin volume: 20 to 500μL
- Filter type: polyethylene filter, 30μm (20 to 50μm) pore size
- Cap type: snap cap on column (no cap on collection tube); press-on bottom caps
Micro-Spin Columns (Part No. 89879):
- Column volume: 400μL
- Resin volume: 5 to 100μL
- Filter type: polyethylene filter, 30μm (20 to 50μm) pore size
- Cap type: O-ring screw top caps; press-on bottom caps
Microcentrifuge Tubes (Part No. 69715):
- 2 mL capacity
- Dolphin-style bottom with attached snap cap
- Included and compatible with Part No. 69700
Microcentrifuge Tubes (Part No. 69720):
- 2 mL capacity
- Short, conical bottom with attached snap cap
- Included and compatible with Part No. 69702
Specifications
Specifications
Product Line | Pierce™ |
Quantity | 50 Columns |
Type | Spin Column |
Format | Microcentrifuge, Spin Column |
Column Type | Empty Column, Polyethylene Filter |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.